Micro capillaries, disposable, Kimble®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SP71900-10 SP71900-5 SP71900-50
SCERSP71900-10EA 705 EUR InStock
SCERSP71900-10 SCERSP71900-5 SCERSP71900-50
Micro capillaries, disposable, Kimble®
Pipetten Mikrokapillaren
These disposable micro capillary pipettes are made from soda lime glass, USP Type III.
  • Calibrated to contain
  • Colour-coded
  • Marked with single black capacity ring
  • Packaged in easy-open cylinders for convenient dispensing
Zertifizierungen: Designed from ASTM Specification E672.
Verpackung: Each cylinder contains 250 pipettes and one pipette device.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge