Culture tubes, disposable, KIMBLE®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SP73500-25150 SP73500-18150 SP73500-16100A
392-0368EA 311 EUR InStock
392-0368 392-0367 SCERSP73500-16100A
Culture tubes, disposable, KIMBLE®
Röhrchen Kulturröhrchen
Culture tubes made from 51 expansion borosilicate glass that offer excellent chemical resistance against sodium leaching, a factor common in soda lime glass products.
  • Features sturdy, uniform bottoms and consistent lengths
  • Suitable for cell washing procedures
  • Plain and disposable
Zertifizierungen: Glass meets ASTM E438, Type I, Class B requirements. Designed from ASTM E890 specifications.
Verpackung: Tubes are packed in convenient trays that are shrink-wrapped in plastic film, with tubes remaining in constant alignment for protection against in-transit breakage and contamination.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge