Tissue grinders, Tenbroeck type, KIMBLE®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SP885000-0040 SP885000-0015 SP885000-0002 SP885000-0007
SCERSP885000-0040EA 467 EUR InStock
SCERSP885000-0040 SCERSP885000-0015 SCERSP885000-0002 SCERSP885000-0007
Tissue grinders, Tenbroeck type, KIMBLE®
Homogenisatoren Gewebehomogenisatoren
Borosilicate glass with ground glass pestle.
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • For hand or motor operation
  • Replacement parts are completely interchangable
  • Clearance between pestles and tubes is 0,10 to 0,15 mm
The pestle is tooled for a single hole rubber stopper. By inserting one end of a short metal rod in the stopper, these grinders can be motor driven at slow speeds by inserting the other end of the metal rod into the motor chuck. Ideal for grinding tough connective tissue and soft tissue such as brain or liver.
Bestellinformation: Supplied without rubber stoppers and metal rods (must be ordered separately).
Verpackung: Packaged in a convenient high-density polyethylene case with die cut foam inserts that protect grinders in transit and in storage.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge