Tissue grinders, DUALL®, KIMBLE®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SP885450-0020 SP885450-0021 SP885450-0022 SP885450-0023 SP885450-0024 SP885450-0025
SCERSP885450-0020EA 409 EUR InStock
SCERSP885450-0020 SCERSP885450-0021 SCERSP885450-0022 SCERSP885450-0023 SCERSP885450-0024 SCERSP885450-0025
Tissue grinders, DUALL®, KIMBLE®
Homogenisatoren Gewebehomogenisatoren
These borosilicate glass tissue grinders combine both conical and cylindrical surfaces to reduce tissue and also produces a uniform homogenate. Construction is strong enough to allow the pestle to be motor driven and also the grinding efficiency is greatly improved while using this tube.
  • Replacement components are available and completely interchangeable
  • Used for connective tissues such as muscle, heart and lung
Clearance between pestles and tubes is 0,004 to 0,006"
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge