Tissue grinders, DUALL®, KIMBLE®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SP885480-0020 SP885480-0022 SP885480-0023
SCERSP885480-0020EA 325 EUR InStock
SCERSP885480-0020 SCERSP885480-0022 SCERSP885480-0023
Tissue grinders, DUALL®, KIMBLE®
Homogenisatoren Gewebehomogenisatoren
These tissue grinders (borosilicate glass, PTFE pestle with glass tube) combine both conical and cylindrical surfaces to reduce tissue and also produces a uniform homogenate. Construction is strong enough to allow the pestle to be motor driven and also the grinding efficiency is greatly improved while using this tube.
  • Replacement components are available and completely interchangeable
  • Used for soft tissue such as brain and liver
Clearance between pestles and tubes is 0,004 to 0,006"
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge