Fastpek Allergen ELISA Kits
Lieferant: HYSERVE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1500005 1500007-2
SCIL1500005EA 351 EUR InStock
SCIL1500005 SCIL1500007-2
Fastpek Allergen ELISA Kits
Klinisches Diagnosezubehör Klinische Reagenzien und Kits
These kits are designed to detect allergens by special extraction buffer and antibodies.
  • High sensitivity
  • Increased specificity
  • Assay time of about 120 minutes
Suitable for use in highly processed food.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge