Hydrometers in plastic
Lieferant: Ludwig Schneider GmbH & CO. KG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
3001530 3000015 3007590 3006075 3004560 3003045 3002595 3002523
SCNL3001530EA 130 EUR InStock
SCNL3001530 SCNL3000015 SCNL3007590 SCNL3006075 SCNL3004560 SCNL3003045 SCNL3002595 SCNL3002523
Hydrometers in plastic
Aräometer
High quality polycarbonate hydrometers for %mas/°Brix measures.
  • Breakage protected (no glass breakage)
  • High transparency with easy-to-read scales
  • High precision and reproducibility
Suitable for applications in food, pharmaceutical and cosmetic industries.
Reference temperature: 20 °C
Bestellinformation: Available with works calibration certificate on request.
Verpackung: In square plastic sleeve with individual serial number.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge