Diffusor for ring light, VisiLED, S40-75
Lieferant: SCHOTT AG Lighting and imaging
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
400580
SCOC400580EA 311 EUR InStock
SCOC400580
Diffusor for ring light, VisiLED, S40-75
Beleuchtung Ringlichter
The diffusor for LED ring lights avoids reflections of LEDs for uniform illumination.
  • Diffusor with adapting thread (M110×1) to avoid reflections of LEDs
Designed for use only with VisiLED ringlight S40-75.
Bestellinformation: VisiLED ringlight S40-75 and controller MC 750, MC 1100, or MC 1500 required.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge