Darkfield optic ring for VisiLED slim ring light
Lieferant: SCHOTT AG Lighting and imaging
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
400810
SCOC400810EA 223 EUR InStock
SCOC400810
Darkfield optic ring for VisiLED slim ring light
Beleuchtung Ringlichter
Focus optic ring for darkfield illumination.
  • Focus optics ring for darkfield
  • Working distance: 5 to 25 mm
Designed for use only with VisiLED slim ring light (SCOC400150).
Bestellinformation: VisiLED slim ring light and controller MC 750, MC 1100, or MC 1500 required.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge