Electrolyte solutions, organic
Lieferant: SI Analytics
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
285138308 285138324
SCOI285138308EA 80 EUR InStock
SCOI285138308 SCOI285138324
Electrolyte solutions, organic
Elektrolyte Elektrodenfülllösungen
Electrolyte solutions for measurements in organic solutions for reference electrodes and as electrolyte bridges.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge