Cannon-Fenske viscometers for manual measurements
Lieferant: SI Analytics
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
285403523 285403507 285403515 285403531 285403548 285403556 285403564 285403572 285403589 285403597 285403601 285403618
SCOI285403523EA 297 EUR InStock
SCOI285403523 SCOI285403507 SCOI285403515 SCOI285403531 SCOI285403548 SCOI285403556 SCOI285403564 SCOI285403572 SCOI285403589 SCOI285403597 SCOI285403601 SCOI285403618
Cannon-Fenske viscometers for manual measurements
Viskosimeter Kapillarviskosimeter
Cannon-Fenske viscometers for manual measurements especially in routine analyses.
Suitable for transparent liquids, foaming liquids and liquid mixture with highly volatile components.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge