Cannon-Fenske viscometers for automatic measurements
Lieferant: SI Analytics
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
285403712 285403729 285403745 285403753 285403761 285403778 285403786 285403794 285403807 285403737 285403815
SCOI285403712EA 297 EUR InStock
SCOI285403712 SCOI285403729 SCOI285403745 SCOI285403753 SCOI285403761 SCOI285403778 SCOI285403786 SCOI285403794 SCOI285403807 SCOI285403737 SCOI285403815
Cannon-Fenske viscometers for automatic measurements
Viskosimeter Kapillarviskosimeter
Cannon-Fenske viscometers for automatic measurements especially in routine analyses.
Suitable for transparent liquids, foaming liquids, and liquid mixture with highly volatile components.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge