Flow through cooler CK 310
Lieferant: SI Analytics
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
285414320
SCOI285414320EA 4790 EUR InStock
SCOI285414320
Flow through cooler CK 310
Umwälzthermostate Geschlossene Umwälzsysteme Kälteumwälzthermostate
The flow-through cooler CK 310 is the perfect complement to bath thermostats of CT 72 series to extending their temperature range to below ambient temperature.
  • Small footprint
  • Low energy requirement: Economic solution preventing tap water usage for cooling
  • Internal cooling loop is made of stainless steel: Better chemical resistance than copper
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge