DURAN® Glass Tubes
Lieferant: Schott AG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
DR19/1.9AR DR38/2.0AR 1028437 DR40/2.4AR 322.0AR 1485594 1544262 1544261 1552469 1580606 1580607 1580608 1580609 1621123 1637429 1637432 1397497 1529018 1662270 1552466 1028561 1667897 1643569 1716790 1716791 1716799 1716852 1027516 1027813
SCORDR19/1.9AREA 0 EUR InStock
SCORDR19/1.9AR SCORDR38/2.0AR SCOR1028437 SCORDR40/2.4AR SCOR322.0AR SCOR1485594 SCOR1544262 SCOR1544261 SCOR1552469 SCOR1580606 SCOR1580607 SCOR1580608 SCOR1580609 SCOR1621123 SCOR1637429 SCOR1637432 SCOR1397497 SCOR1529018 SCOR1662270 SCOR1552466 SCOR1028561 SCOR1667897 SCOR1643569 SCOR1716790 SCOR1716791 SCOR1716799 SCOR1716852 SCOR1027516 SCOR1027813
DURAN® Glass Tubes
Schläuche Transferschlauch
High precision glass tubes made from AR® glass.
  • Good chemical and physical properties
  • Not resistant to high thermal loads or rapid temperature changes
DURAN® Glass tubes are suitable for the production of items that must normally withstand only brief exposure to aggressive chemicals.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge