Flasche, Enghals, mit Normschliff, DURAN®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
211647301 211648606 211649105 211643608 211642406 211644407 211645403 211646305 211641701
SCOT211647301EA 215 EUR InStock
SCOT211647301 SCOT211648606 SCOT211649105 SCOT211643608 SCOT211642406 SCOT211644407 SCOT211645403 SCOT211646305 SCOT211641701
Flasche, Enghals, mit Normschliff, DURAN®
Flaschen Glasflaschen
DURAN®, borosilicate glass 3.3, clear or brown, with standard ground joint, without stopper.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP
Brown tinted to protect light-sensitive substances.
With printed retrace code (batch identification).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge