Flasche, Weithals, mit Normschliff, DURAN®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
211847306 211848602 211849101 211845408 211844403 211846301 211842402 211843604
SCOT211847306EA 241 EUR InStock
SCOT211847306 SCOT211848602 SCOT211849101 SCOT211845408 SCOT211844403 SCOT211846301 SCOT211842402 SCOT211843604
Flasche, Weithals, mit Normschliff, DURAN®
Flaschen Glasflaschen
DURAN®, borosilicate glass 3.3, clear or amber, with standard ground joint.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP
Without stopper. Marked with volume and equipped with a white labelling field. Amber to protect light-sensitive materials.
With printed retrace code (batch identification).
Zertifizierungen: ISO 4796-2
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge