Sedimentiergefäße, Imhoff, DURAN®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
214015403
SCOT214015403EA 110 EUR InStock
SCOT214015403
Sedimentiergefäße, Imhoff, DURAN®
Trichter Sedimentiergefäße
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • Very good chemical resistance
  • Without stopcock
The conical sedimentation cone with graduation according to Imhoff allows a quantification of sediments from a suspension (e.g. in waste water).
Zertifizierungen: According to DIN 12672.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge