Measuring Pipettes, Class AS
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
233470607 233471106 233471603 233472308 233472908 233473201 233473407 233473604 233490609 233491108 233491605 233492301 233492901 233493203 233493409 233493606
SCOT233470607EA 94.6 EUR InStock
SCOT233470607 SCOT233471106 SCOT233471603 SCOT233472308 SCOT233472908 SCOT233473201 SCOT233473407 SCOT233473604 SCOT233490609 SCOT233491108 SCOT233491605 SCOT233492301 SCOT233492901 SCOT233493203 SCOT233493409 SCOT233493606
Measuring Pipettes, Class AS
Pipetten Messpipetten (Mohr)
Pipettes are used for precise measurement and decanting of liquids. Graduated pipettes allow different amounts of liquid to be absorbed and dispensed in the same or in different sizes. The tolerances for the volume content of calibratable pipettes correspond to the error limits of the German weights and measures regulations as well as ISO and DIN.
  • Manufactured from soda-lime glass
  • Rapid discharge reduces significantly the waiting time compared to Class A
  • Calibration is based on the poured out volume ('Ex') at a +20 °C reference temperature
  • With circular ring graduations, with cotton plug
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable amber/blue print
  • Waiting time of 15 seconds
  • Autoclavable
The blow-out allows different quantities of liquids to be held and dispensed in portions of the same size. The amber/blue scale is provided with main point ring division.
Amber print: Type 2 - blow-out, nominal volume at top (Numbering from bottom to top)
Blue print: Type 3 - blow-out, zero at top (Numbering from the top down)
Zertifizierungen: Complies with ISO 835.
Lieferumfang: Pipettes with a batch number and accuracy Class AS are supplied with a batch certificate.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge