Planschliff-Glocke, mit Normschliff-Hals, DURAN®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
244655907 244656106 244656903
SCOT244655907EA 250 EUR InStock
SCOT244655907 SCOT244656106 SCOT244656903
Planschliff-Glocke, mit Normschliff-Hals, DURAN®
Dosen Glocken
DURAN®, borosilicate glass 3.3, clear.
  • Good chemical and physical properties
  • Very good thermal resistance
  • Quick and easy to clean
Wall thickness and geometry designed to suit vacuum applications.
The flat flange bell is used for tests with a vacuum experimental plate (vacuum chamber). The bell has a ground flange and a ground joint for the insertion of measuring equipment.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge