Brushes, for NMR tubes, DURAN®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
290740059
SCOT290740059EA 3.3 EUR InStock
SCOT290740059
Brushes, for NMR tubes, DURAN®
Bürsten
Stubborn soiling and deposits in NMR tubes can be easily removed with the NMR brush.
  • Due to the accuracy of fit, bristles rest against the inner wall of the glass for an optimal cleaning process
  • Recommended usable temperature range is –70 to 200 °C
  • Stainless steel wire with nylon bristles, curved end of bristle trim
Nuclear magnetic resonance spectroscopy (NMR-spectroscopy) is a spectroscopic method to elucidate the structure of molecules and compounds. This analytical method requires very precise tubes with a constant diameter, a uniform wall thickness, and excellent straightness across the entire tube. Deposits and contamination in NMR tubes can be dissolved with the help of the NMR brush. Subsequent cleaning with a suitable solvent completely acidifies the NMR tube.
Max. operating temperature: 200 °C
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge