Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Rubber stopper, DURAN®

Lieferant: DWK Life Sciences

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

292062803 292061301
215-4191EA 33.9 EUR InStock
215-4191 SCOT292061301
Rubber stopper, DURAN®
Stopfen Gummistopfen
This bromobutyl rubber closure provides a gas-tight seal for all GL 25 and GL 45 laboratory bottles with sizes from 10 to 20000 ml.

  • Elastic material for reliable self-sealing after needle piercing
  • Upper surface features seven numbered hemispherical impressions for needle piercing
  • Resistant to temperature from –50 to +121 °C

Suitable for storing oxygen-sensitive materials, cultivation of anaerobic microorganisms (pressure resistent bottles recommended), aerobic methane oxidation (MOx) studies, storage of anaerobic culture media (amber glass bottles recommended), applications requiring gas purging or sample transfer through syringes.

Zertifizierungen: Conforms to EP 3.2.9 Type I closure for chemical and penetration tests.

Achtung! Bitte beachten: Standard DURAN® original GL 45 lab bottles are not pressure resistant. If build up in pressure is anticipated, it is recommended to use the stopper without the GL 45 red open top cap. The use of pressure resistant bottles such as DURAN® pressure plus+ should be considered.
Needles larger than 20 gauge are not recommended.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge