PTFE coated silicone seals, DURAN®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
292489102 292481456 292482958
SCOT292489102EA 92.2 EUR InStock
SCOT292489102 SCOT292481456 SCOT292482958
PTFE coated silicone seals, DURAN®
Septen
High-quality silicone seals developed for the needs of the pharmaceutical and biotech industry.
  • Cleanroom packaged, supplied with Certificate of Analysis (CoA), USP <88> Class VI Pharmaceutical grade materials
  • Recommended usable temperature range is –196 to 200 °C
  • Sterilisable via steam/autoclave, chemical, or dry heat
  • Lab washer compatible and microwave safe
High-quality, PTFE coated silicone seals are designed for use with DURAN® PURE Premium Screw Caps. The silicone portion is light ivory in color, while the PTFE portion is gray.
Verpackung: 10 pieces supplied per bag.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge