HighFlow Solution System: - XL funnel with lid, OD 200 mm, Material: PE-HD-EL, including coarse sieve, Thread: male thread M34x2 - Main pipe, OD 24 mm, ID 20 mm, length approx. 800 mm (Material: stainless steel 1.4571 (V4A), Thread: female thread M34 1 * 1 ST
Lieferant: SCAT VERTRIEB
HighFlow Solution System: - XL funnel with lid, OD 200 mm, Material: PE-HD-EL, including coarse sieve, Thread: male thread M34x2 - Main pipe, OD 24 mm, ID 20 mm, length approx. 800 mm (Material: stainless steel 1.4571 (V4A), Thread: female thread M34 1 * 1 ST
New Product
PRODUKT
Erweitert / Kompakt
HighFlow Solution System: - XL funnel with lid, OD 200 mm, Material: PE-HD-EL, including coarse sieve, Thread: male thread M34x2 - Main pipe, OD 24 mm, ID 20 mm, length approx. 800 mm (Material: stainless steel 1.4571 (V4A), Thread: female thread M34 1 * 1 ST SCTV106623 1 * 1 ST
New Product
Hersteller-Artikelnummer:: 106623
Lokale Artikelnummer:: SCTV106623
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder