Accessories for SCAT Politainer
Lieferant: S.C.A.T.
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
107338
SCTV107338EA 64.9 EUR InStock
SCTV107338
Accessories for SCAT Politainer
Sicherheitsbehälter
The SCAT Politainer - also known as folding canisters - is a combination of a flexible plastic bag and a stable UN-approved outer carton. Storage in the unfilled state is possible in a very small space and after filling, safe storage or transport is guaranteed.
  • Thread to adjust the thread from GL38 to GL45
  • Especially suitable for SCAT Politainer
SCAT Politainers are an ideal way of not having to dispense with safe storage even in limited spaces.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge