Open top, 2 or 3 compartment dispensers
Lieferant: S-CURVE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BD-03
SCURBD-03EA 387 EUR InStock
SCURBD-03
Open top, 2 or 3 compartment dispensers
Spender für PSA
Two- and three-compartment dispensers are made from clear, clean, 6 mm thick acrylic.
Open top provides easy access to items. Ideal for gloves, glove liners, face masks, and other disposables. Edges are flame-treated for smoothness. Rubber pads on bottom corners prevent slipping.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge