Baths for immersion thermostat, Termotronic II
Lieferant: SELECTA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
6000391 6000393
SELE6000391EA 577 EUR InStock
SELE6000391 SELE6000393
Baths for immersion thermostat, Termotronic II
Bäder Wasserbäder, ohne Umwälzung
These metal baths are made up of double external AISI 304 stainless steel skin and an AISI 310 stainless steel pressed bath interior.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge