DIM-C-pPhOH Synonyms: C-DIM8, CDIM8 1* 25mg CAS 151358-47-3 DIM-C-pPhOH (C-DIM8, CDIM8) is a nuclear receptor 4A1 (NR4A1)(Nur77) antagonist. DIM-C-pPhOH inhibits tumor growth and mTOR signaling, induces apoptosis and cellular stress. DIM-C-pPhOH redu 1 * 25 mg
Lieferant: SELLECK CHEMICALS
DIM-C-pPhOH Synonyms: C-DIM8, CDIM8 1* 25mg CAS 151358-47-3 DIM-C-pPhOH (C-DIM8, CDIM8) is a nuclear receptor 4A1 (NR4A1)(Nur77) antagonist. DIM-C-pPhOH inhibits tumor growth and mTOR signaling, induces apoptosis and cellular stress. DIM-C-pPhOH redu 1 * 25 mg
PRODUKT
Erweitert / Kompakt
DIM-C-pPhOH Synonyms: C-DIM8, CDIM8 1* 25mg CAS 151358-47-3 DIM-C-pPhOH (C-DIM8, CDIM8) is a nuclear receptor 4A1 (NR4A1)(Nur77) antagonist. DIM-C-pPhOH inhibits tumor growth and mTOR signaling, induces apoptosis and cellular stress. DIM-C-pPhOH redu 1 * 25 mg SELKS6799-25MG 1 * 25 mg
Hersteller-Artikelnummer:: S6799-25MG
Lokale Artikelnummer:: SELKS6799-25MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder