Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Recombinant Human Glutathione Peroxidase 2

Lieferant: SELENOZYME

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


HGPX2-05 HGPX2-1 HGPX2-2.5 HGPX2-5
SELZHGPX2-05EA 450 EUR InStock
SELZHGPX2-05 SELZHGPX2-1 SELZHGPX2-2.5 SELZHGPX2-5
Recombinant Human Glutathione Peroxidase 2
Proteine und Peptide
The human GPX2 selenoprotein is encoded by the GPX2 gene and has potent hydrogen peroxide reducing activity using glutathione.
We provide the human GPX2 enzyme with approx. 20% Sec contents, mixed with inactive forms containing Sec-to-Lys and Sec-to-Gln substitutions, at more than 95% purity in aliquots at amounts from 0,5 to 5 units.

  • Genuine Sec-containing selenoproteins
  • No animals used as protein sources (grown in bacteria)
  • More than 90% Sec contents
  • More than 95% purity

Lieferumfang: Protein stored in 25 mM Tris-HCl (pH 8.0) with 250 mM KCl, 2 mM 2-Mercaptoethanol and 10% glycerol. Concentration 1 mg protein/ml. Shipped in cold pack by courier service. Store at −20 °C upon delivery.

Verpackung: Aliquots

Achtung! Bitte beachten: Centrifuge the tube briefly before opening the lid.


Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge