Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Recombinant Human Thioredoxin Reductase 2

Lieferant: SELENOZYME

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


HTRXR2-5 HTRXR2-10 HTRXR2-20 HTRXR2-50
SELZHTRXR2-5EA 750 EUR InStock
SELZHTRXR2-5 SELZHTRXR2-10 SELZHTRXR2-20 SELZHTRXR2-50
Recombinant Human Thioredoxin Reductase 2
Proteine und Peptide
The human TrxR2 selenoprotein is encoded by the TXNRD2 gene and exerts a wide range of functions in mitochondrial redox control and antioxidant systems. We provide the main mitochondrial isoform of the human TrxR2 enzyme with more than 90% Sec contents and more than 95% purity in aliquots at amounts from 5 to 50 units.

  • Genuine Sec-containing selenoproteins
  • No animals used as protein sources (grown in bacteria)
  • More than 90% Sec contents
  • More than 95% purity

Lieferumfang: Protein stored in TE buffer with 50% glycerol. Concentration 1 mg protein/ml. Shipped in cold pack by courier service. Store at −20 °C upon delivery.

Verpackung: Aliquots

Achtung! Bitte beachten: Centrifuge the tube briefly before opening the lid.


Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge