Pulvertrichter
Lieferant: SEMADENI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0211 0213
SEMA0211EA 9.2 EUR InStock
SEMA0211 SEMA0213
Pulvertrichter
Trichter Pulvertrichter
Funnels, made of PP, translucent. Used for transferring powders. Also suitable for liquids.
  • Parallel stem minimises powder bridging
  • Very good chemical and temperature resistance
  • Autoclavable
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge