Semi-micro cuvette
Lieferant: SEMADENI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
12601
SEMA12601EA 0 EUR InStock
SEMA12601
Semi-micro cuvette
Küvetten Spektralphotometer-Küvetten
Cuvette for spectrophotometer with the lowest tolerances.
  • Round opening
  • Excellent transparency and optical properties
Especially suitable for photometric measurements in the UV range.
Verpackung: Individually protected in Styrofoam boxes.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge