Centrifuge tubes
Lieferant: SEMADENI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1567
SEMA1567EA 5.4 EUR InStock
SEMA1567
Centrifuge tubes
Röhrchen Zentrifugenröhrchen
PP, clear.
  • Stable
  • Autoclavable
  • Temperature resistant up to 130 °C
  • Excellent chemical resistance
Round bottom with rim.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge