Packaging bottles, narrow neck, rectangular, HDPE, with screw caps
Lieferant: SEMADENI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2026 2025 2024
215-0240EA 263 EUR InStock
215-0240 SEMA2025 SEMA2024
Packaging bottles, narrow neck, rectangular, HDPE, with screw caps
Flaschen Kunststoffflaschen
HDPE, translucent, with thread.
  • Good chemical resistance
  • Good temperature resistance
Bestellinformation: Screw caps must be ordered separately.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge