Draining racks, wall mounted
Lieferant: SEMADENI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1557 2110
211-2084EA 342 EUR InStock
211-2084 SEMA2110
Draining racks, wall mounted
Gestelle Abtropfgestelle
White, PVC drain channel, polyethylene coated metal rack.
  • Drain channels inclined toward the centre and provided with a drain point
  • Metal frame and drip plasticised PE rods
  • The rack is hung on the wall plate and can be easily removed for cleaning
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge