Containers, disposable, with cap
Lieferant: SEMADENI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2988
SEMA2988EA 42.3 EUR InStock
SEMA2988
Containers, disposable, with cap
Probenbehälter Universalbehälter
Transparent cup made of PS with well fitting PE cap, universally applicable.
  • Good chemical resistance
  • Good temperature resistance
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge