Desiccator cabinet, horizontal, PMMA
Lieferant: SEMADENI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
3463
SEMA3463EA 0 EUR InStock
SEMA3463
Desiccator cabinet, horizontal, PMMA
Exsikkatoren Trockenkammern
Transparent PMMA cabinet that has white ABS frame and transparent PVC sides. Ideal for storing moisture sensitive samples or electronic components. Unit is not suitable for use under vacuum or pressure.
  • Door mounted hygrometer is graduated from 0 to 100%
  • Cabinet has two lateral interlocking gas connectors. Check valves open automatically and close when hose is disconnected, allowing gas to escape
Lieferumfang: Supplied with 2 removable acrylic glass shelves.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge