Jars, with screw cap
Lieferant: SEMADENI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
3721_U 3720 3715 3716 5241 3714 3718
SEMA3721_UEA 29.6 EUR InStock
SEMA3721_U SEMA3720 SEMA3715 SEMA3716 SEMA5241 SEMA3714 SEMA3718
Jars, with screw cap
Probenbehälter Universalbehälter
PP, white.
  • Good chemical resistance
  • Good temperature resistance
Ideal for collecting, storing and transporting samples.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge