Staining tray
Lieferant: SEMADENI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
4332
SEMA4332EA 67.6 EUR InStock
SEMA4332
Staining tray
Färbebehälter
Transparent base with a well fitting PP lid.
  • Usable up to 70 °C
  • Stackable with two handles
  • Good chemical resistance
Suitable for dying, bleaching and fixing of gels and membranes susceptible electrophoresis.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge