Storage cabinet
Lieferant: SEMADENI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
5187
SEMA5187EA 43.3 EUR InStock
SEMA5187
Storage cabinet
Möbel Lagerschränke
The storage cabinet system allows a quick, individual and efficient division of drawers, shelves, cupboards, etc.
  • The boxes can be put together in a number of variations according to a sophisticated module
  • The boxes have holders on the bottom, which fit into the grid area of ​​the self-adhesive drawer foil
  • This self-adhesive foil prevents the boxes from slipping around in the drawer
  • Heavy-duty boxes made of impact-resistant polystyrene (PS).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge