Storage Boxes with Lid
Lieferant: SEMADENI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
7256 7258_U 7251 7246 7247 7254 7255
SEMA7256EA 13.1 EUR InStock
SEMA7256 SEMA7258_U SEMA7251 SEMA7246 SEMA7247 SEMA7254 SEMA7255
Storage Boxes with Lid
Boxen Lagerboxen
These fresh storage boxes made of polypropylene (PP) are dimensionally stable.
  • With hermetically closing blue snap-lid made of soft polythene (PE-LD)
  • Can be used for temperatures from −40 to +85 °C
  • Dishwasher and microwave resistant
  • Approved for food contact
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge