Module for AiroSensor X ER 20-20-43, Adapter Cable for Customer Supplied Pt100
Lieferant: SenseAnywhere
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0096647494838
SENA0096647494838EA 65 EUR InStock
SENA0096647494838
Module for AiroSensor X ER 20-20-43, Adapter Cable for Customer Supplied Pt100
Datenlogger
This smart probe interface featuring a digital SenseAnywhere Bus interface has been especially designed to connect an existing 4-wire Pt100 Probe to the SenseAnywhere Monitoring System.
  • Sensor interface for existing 4-wire Pt100 (3-wire Pt100 not supported)
Due to the digital interface this module with its own serial number and integrated electronics can be calibrated. Calibrated SAB modules can be easily hot swapped offering a big advantage and cost savings for recalibration of sensors.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge