OPTI-LYNX™ Micro Trap Cartridges
Lieferant: SEQUANT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
11-04755-TD
SEQT11-04755-TDEA 0 EUR InStock
SEQT11-04755-TD
OPTI-LYNX™ Micro Trap Cartridges
Chromatographiesäulen
Combining a versatile selection of packed beds and the use of a convenient quick-connect holder, OPTI-LYNX™ provides numerous options for chromatographers for on-line or off-line sample clean-up and pre-concentration.
  • Quick-connect system in-line (BC) or direct-connect versions
  • 414 bar with hand-tight quarter turn, no tools needed
  • Low back pressure design
  • Bi-directional
  • Biocompatible cartridges
  • Available in packages of five cartridges
Two styles of holders are available: The Opti-Lynx Micro BC offers full biocompatibility as the in-line version while the direct-connect configuration threads conveniently into all 10-32 ports.
OPTI-LYNX™ traps are the ideal tool for optimising your technique, whether you want to separate a peptide digest from its matrix for further analyses, or prepare a dilute small molecule sample for LC injection without loss of sample. These columns may be loaded and regenerated repeatedly for maximum value.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge