Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Silikonelastomer, Premium Care MED-4086

Lieferant: SF MEDICAL

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

MED-4086-SH3MM
SFAAMED-4086-SH3MMEA 0 EUR InStock
SFAAMED-4086-SH3MM
Silikonelastomer, Premium Care MED-4086
Silikonelastomer
A very soft two-part, low viscosity, silicone elastomer for use in potting, encapsulating, coating, and injection or transfer molding application.

  • Cures with heat via addition-cure chemistry
  • Ratio of 1:1 mix (Part A: Part B)

The properties (specifications) are tested on a lot-to-lot basis. Do not use as a basis for preparing specifications. Please contact NuSil Technology for assistance and recommendations in establishing particular specifications


Order Now
Spezifikation Testergebnisse
Durometer 55 Type 000
Viscosity 6,450 cP
Work Time 18 hours
Tensile 40 psi
Cure 45 minutes / 150°C
Cure System Platinum
Elongation 4,75
Mix Ratio 1:1
Stress @ Strain 8 psi / 200 %
Comment Ultra Soft Elastomer

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge