On-column syringes
Lieferant: SGE Analytical Science
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
000376 000380
SGEA000376EA 141.3 EUR InStock
SGEA000376 SGEA000380
On-column syringes
Spritzen Spritzen für die Chromatographie
On-column syringes are suitable for general purpose applications.
When selecting an on-column syringe, the needle outside diameter (OD) must be smaller than the inside diameter (ID) of the capillary column it will be used with.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge