Precision bore PEEK tubing is designed specifically for modern LC, LCMS and automation usage.
- Chemical resistant and inert to most commonly used solvents
- Bio-inert and bio-compatible
- Colour coded to industry standards for easy identification of inner diameter
- Leach-free colorants
- Smooth inner walls and precise IDs
- Works well with polymer and stainless steel fittings
- Maximum operating temperature of 100 °C
PEEK tubing is a flexible alternative to stainless steel tubing in high pressure applications. Also ensures reproducibility in chromatographic flow rates through smooth internal surface along with consistent and tight toleranced IDs.
Bestellinformation: Accessories should be ordered separately.