CRS SPure™ H₂O/HC Filters
Lieferant: SGE Analytical Science
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
202324-4-PC 202324-6-PC 202324-B 202324-B-QC 202324D-4-PC 202324D-6-PC 202324D-B 202324D-B-QC 202324DRP-B-QC 202324DRP-SS-Q 202324D-SS 202324D-SS-QC 202324L-4-PC 202324L-6-PC 202324L-B 202324L-B-QC 202324LRP-B-QC 202324LRP-SS-Q 202324L-SS 202324L-SS-QC 202324RP-B-QC 202324RP-SS-QC 202324-SS 202324-SS-QC 202324XL-4-PC 202324XL-6-PC 202324XL-B 202324XL-B-QC 202324XLRP-B-Q 202324XLRP-SS- 202324XL-SS 202324XL-SS-QC 202325-B 202325-B-QC 202325D-B 202325D-B-QC 202325D-SS 202325D-SS-QC 202325L-B 202325L-B-QC 202325L-SS 202325L-SS-QC 202325-SS 202325-SS-QC 202325XL-B 202325XL-B-QC 202325XL-SS 202325XL-SS-QC
SGEA202324-4-PCEA 137.1 EUR InStock
SGEA202324-4-PC SGEA202324-6-PC SGEA202324-B SGEA202324-B-QC SGEA202324D-4-PC SGEA202324D-6-PC SGEA202324D-B SGEA202324D-B-QC SGEA202324DRP-B-QC SGEA202324DRP-SS-Q SGEA202324D-SS SGEA202324D-SS-QC SGEA202324L-4-PC SGEA202324L-6-PC SGEA202324L-B SGEA202324L-B-QC SGEA202324LRP-B-QC SGEA202324LRP-SS-Q SGEA202324L-SS SGEA202324L-SS-QC SGEA202324RP-B-QC SGEA202324RP-SS-QC SGEA202324-SS SGEA202324-SS-QC SGEA202324XL-4-PC SGEA202324XL-6-PC SGEA202324XL-B SGEA202324XL-B-QC SGEA202324XLRP-B-Q SGEA202324XLRP-SS- SGEA202324XL-SS SGEA202324XL-SS-QC SGEA202325-B SGEA202325-B-QC SGEA202325D-B SGEA202325D-B-QC SGEA202325D-SS SGEA202325D-SS-QC SGEA202325L-B SGEA202325L-B-QC SGEA202325L-SS SGEA202325L-SS-QC SGEA202325-SS SGEA202325-SS-QC SGEA202325XL-B SGEA202325XL-B-QC SGEA202325XL-SS SGEA202325XL-SS-QC
CRS SPure™ H₂O/HC Filters
Filter Gasfilter
The SPure H₂O/HC Filter removes moisture, oil, dust, and hydrocarbons (C5 and heavier) from dry air and inert gases.
The cobalt-free moisture indicator signals breakthrough at 5% relative humidity and changes from green to blue.
SPure Filters have the option of standard compression fittings and quick connect fittings in ¹/₄ and ¹/₈" brass and stainless steel, or metal push-to-connect fittings in ¹/₄" and 6 mm for plastic tubing and non-carrier gas lines.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge