Pure-Pass™ Sealing Mats for Well Plates
Lieferant: SGE Analytical Science
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
462001 465001 465002 461001 461010
SGEA462001EA 37.4 EUR InStock
SGEA462001 SGEA465001 SGEA465002 SGEA461001 SGEA461010
Pure-Pass™ Sealing Mats for Well Plates
Versiegeln Mikrotiterplatten-Dichtungen
Pure-Pass sealing mats protect wells from leaks, contamination and evaporation during sampling and storage.
  • Available in non-slit and slit versions
  • Slit version enhances sample recovery by reducing vacuum formation
  • Plasma coating reduces the natural tackiness of silicone
These mats are available in EVA and specially processed low bleed, low extractable silicone to reduce sticking and a pre-slit version is available for easier piercing and sample withdrawal.
All mats are for 96 well plates with 7 mm diameter wells. Excellent dimensional conformance with ANSI/SLAS ensures enhanced registration with the plate for ease of application. Each mat has an alphanumeric grid reference.
Pure-Pass EVA sealing mats are clean room molded. These have dome shape plugs to ease insertion and thin web eases piercing.
Pure-Pass silicone sealing mats offers the excellent chemical resistance of silicone. These are specially processed to reduce risk of accidental sample contamination (low bleed/extractables).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge