Fetal bovine serum (United States), Hyclone™, standard
Lieferant: GE Healthcare - Hyclone
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SH30088.03IH25
SH30088.03IH25EA 1010 EUR InStock
SH30088.03IH25
Fetal bovine serum (United States), Hyclone™, standard
Seren
HyClone standard fetal bovine serum (FBS), U.S. origin is an economical alternative to characterised and defined FBS. Standard FBS is ideal for research applications.
  • Sourced in the USA with complete traceability back to original source
  • Low in antibodies and high in growth factors
  • Produced using true pool technology for uniformity and consistency from bottle to bottle
  • Filtered through three sequential 100 nm (0.1 µm) pore size-rated filters
  • Virus panel testing according to 9 CFR 113.53
Standard FBS is assayed for gamma globulin, alkaline phosphatase, lactate dehydrogenase, glutamic pyruvic transaminase, glutamic oxaloacetic transaminase, pH, total protein, albumin, blood urea nitrogen, creatinine, total bilirubin, sodium, potassium, calcium, chloride, inorganic phosphorus, osmolality, iron, total iron binding capacity (TIBC) and IgG. Included assays are subject to change without notice.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge