HyClone™ Fetal Bovine Serum (United States), Super Low IgG, HyClone Products (Cytiva)
Lieferant: GE Healthcare - Hyclone
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SH30898.02 SH30898.03 SH30898.02HI SH30898.03HI SH30898.03IH25 SH30898.03IR25 SH30898.01
HYCLSH30898.02EA 326 EUR InStock
HYCLSH30898.02 HYCLSH30898.03 HYCLSH30898.02HI HYCLSH30898.03HI SH30898.03IH25 SH30898.03IR25 SH30898.01
HyClone™ Fetal Bovine Serum (United States), Super Low IgG, HyClone Products (Cytiva)
Seren
Enhance cell culture growth with U.S.-sourced Cytiva HyClone Super Low IgG Fetal Bovine Serum (FBS).
  • Processed via protein G chromatography
  • Enhance cell culture growth
  • U.S. sourced
Enhance cell culture growth with U.S.-sourced Cytiva HyClone Super Low IgG Fetal Bovine Serum (FBS), processed via protein G chromatography to substantially reduce immunoglobulin G (IgG) levels.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge