Cell culture medium, HyClone™ ActiPro
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SH31037.01 SH31039.04 SH31037.08 SH31039.03
SH31037.01EA 506 EUR InStock
SH31037.01 SH31039.04 SH31037.08 SH31039.03
Cell culture medium, HyClone™ ActiPro
Zellkulturmedien
HyClone™ ActiPro cell culture medium has been formulated to provide high yields of recombinant proteins in processes using Chinese hamster ovary (CHO) cell lines.
  • Demonstrated high yields of recombinant proteins
  • Does not contain hypoxanthine and thymidine
  • Chemically defined ADCF formulation
  • Production medium for batch or fed-batch processes
  • Accompanied with detailed protocols for use to maximise output
HyClone™ ActiPro does not contain any growth factors such as insulin, peptides, hydrolysates, phenol red, or 2-mercaptoethanol, ensuring batch-to-batch consistency and increased cell culture process efficiency.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge